Merck
  • Metabolomic adaptations and correlates of survival to immune checkpoint blockade.
Metabolomic adaptations and correlates of survival to immune checkpoint blockade.
Nature communications (2019-09-27)
Haoxin Li, Kevin Bullock, Carino Gurjao, David Braun, Sachet A Shukla, Dominick Bossé, Aly-Khan A Lalani, Shuba Gopal, Chelsea Jin, Christine Horak, Megan Wind-Rotolo, Sabina Signoretti, David F McDermott, Gordon J Freeman, Eliezer M Van Allen, Stuart L Schreiber, F Stephen Hodi, William R Sellers, Levi A Garraway, Clary B Clish, Toni K Choueiri, Marios Giannakis
ABSTRACT
Despite remarkable success of immune checkpoint inhibitors, the majority of cancer patients have yet to receive durable benefits. Here, in order to investigate the metabolic alterations in response to immune checkpoint blockade, we comprehensively profile serum metabolites in advanced melanoma and renal cell carcinoma patients treated with nivolumab, an antibody against programmed cell death protein 1 (PD1). We identify serum kynurenine/tryptophan ratio increases as an adaptive resistance mechanism associated with worse overall survival. This advocates for patient stratification and metabolic monitoring in immunotherapy clinical trials including those combining PD1 blockade with indoleamine 2,3-dioxygenase/tryptophan 2,3-dioxygenase   (IDO/TDO) inhibitors.
Sigma-Aldrich
L-Valine-d8, ≥98 atom % D, ≥98% (CP)
Sigma-Aldrich
L-Kynurenine, ≥98% (HPLC)
Sigma-Aldrich
DL-Kynurenine, ≥95.0% (NT)