Merck
  • A small-molecule activator of kinesin-1 drives remodeling of the microtubule network.
A small-molecule activator of kinesin-1 drives remodeling of the microtubule network.
Proceedings of the National Academy of Sciences of the United States of America (2017-12-13)
Thomas S Randall, Yan Y Yip, Daynea J Wallock-Richards, Karin Pfisterer, Anneri Sanger, Weronika Ficek, Roberto A Steiner, Andrew J Beavil, Maddy Parsons, Mark P Dodding
ABSTRACT
The microtubule motor kinesin-1 interacts via its cargo-binding domain with both microtubules and organelles, and hence plays an important role in controlling organelle transport and microtubule dynamics. In the absence of cargo, kinesin-1 is found in an autoinhibited conformation. The molecular basis of how cargo engagement affects the balance between kinesin-1's active and inactive conformations and roles in microtubule dynamics and organelle transport is not well understood. Here we describe the discovery of kinesore, a small molecule that in vitro inhibits kinesin-1 interactions with short linear peptide motifs found in organelle-specific cargo adaptors, yet activates kinesin-1's function of controlling microtubule dynamics in cells, demonstrating that these functions are mechanistically coupled. We establish a proof-of-concept that a microtubule motor-cargo interface and associated autoregulatory mechanism can be manipulated using a small molecule, and define a target for the modulation of microtubule dynamics.
Sigma-Aldrich
Kinesore, >97% (HPLC)