- A simple and expedient method for the preparation of N-chlorohydantoins.
A simple and expedient method for the preparation of N-chlorohydantoins.
Tetrahedron letters (2010-02-17)
Daniel C Whitehead, Richard J Staples, Babak Borhan
PMID20157342
ABSTRACT
A simple and efficient methodology for the preparation of N-chlorinated hydantoins is presented. These versatile chlorenium sources were isolated in high yield after a simple recrystallization. Among the ten examples are the first chiral N-chlorohydantoins.