
Zaleplon Related Compound B

Synonym(s):
N-[3-(3-Cyanopyrazolo[1,5-alpha]pyrimidin-5-yl)phenyl]-N-ethylacetamide
Empirical Formula (Hill Notation):
C17H15N5O
CAS Number:
Molecular Weight:
305.33
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing