
Triptonide

Synonym(s):
14-deoxy-14-oxo-triptolide
Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing