
TOYOPEARL® HW-50F Bulk Media

Synonym(s):
TOYOPEARL® HW-50 Size Exclusion Media
Products
Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing