Skip to Content
Merck
Thiamine mononitrate British Pharmacopoeia (BP) Reference Standard

Thiamine mononitrate

Thiamine mononitrate British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Thiamine nitrate, Vitamin B1 nitrate
Empirical Formula (Hill Notation):
C12H17N5O4S
CAS Number:
Molecular Weight:
327.36
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2