
N-Isobutyryl-L-cysteine

Synonym(s):
N-(2-Methylpropionyl)-L-cysteine
Empirical Formula (Hill Notation):
C7H13NO3S
CAS Number:
Molecular Weight:
191.25
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing