
Homatropine methylbromide for system suitability

Synonym(s):
Homatropine methylbromide, Homatropine methobromide
Empirical Formula (Hill Notation):
C17H24BrNO3
CAS Number:
Molecular Weight:
370.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing