
Ephedrine hydrochloride

Synonym(s):
(1R,2S)-(−)-Ephedrine hydrochloride, (1R,2S)-α-[1-(Methylamino)ethyl]benzyl alcohol hydrochloride, (1R,2S)-(−)-2-Methylamino-1-phenyl-1-propanol hydrochloride
Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH · HCl
CAS Number:
Molecular Weight:
201.69
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2