
Bis(isopropylcyclopentadienyl)tungsten(IV) dihydride

Linear Formula:
(C5H4CH(CH3)2)2WH2
CAS Number:
Molecular Weight:
400.20
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Chemical Composition
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing