
Biacangelicol

Synonym(s):
9-[(3,3-Dimethyl-2-oxiranyl)methoxy]-4-methoxy-7H-furo[3,2-g][1]benzopyran-7-one
Empirical Formula (Hill Notation):
C17H16O6
CAS Number:
Molecular Weight:
316.31
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing