
Apigenin 7-O-neohesperidoside

Synonym(s):
Rhoifolin, Rhoifoloside
Empirical Formula (Hill Notation):
C27H30O14
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing