
Amoxicillin impurity standard

Synonym(s):
Amoxicillin trihydrate
Empirical Formula (Hill Notation):
C16H19N3O5S · 3H2O
CAS Number:
Molecular Weight:
419.45
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing