
Alimemazine for system suitability

Synonym(s):
Alimemazine hemitartrate
Empirical Formula (Hill Notation):
C18H22N2S · 0.5C4H6O6
CAS Number:
Molecular Weight:
373.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing