Skip to Content
Merck
All Photos(2)

Key Documents

155705

Sigma-Aldrich

3-Methoxythiophenol

98%

Synonym(s):

3-Mercaptoanisole, 3-Methoxybenzenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4SH
CAS Number:
Molecular Weight:
140.20
Beilstein:
2041496
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.587 (lit.)

bp

223-226 °C (lit.)

density

1.13 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1cccc(S)c1

InChI

1S/C7H8OS/c1-8-6-3-2-4-7(9)5-6/h2-5,9H,1H3

InChI key

QMVAZEHZOPDGHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Methoxythiophenol was used in the preparation of allenes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contrasting reactivity in Lewis acid-promoted reactions of thio-and silyl-allenes with 1, 4-benzoquinones.
Tetrahedron Letters, 37(3), 327-330 (1996)
Javier Campanini-Salinas et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-22)
A rapid emergence of resistant bacteria is occurring worldwide, endangering the efficacy of antibiotics and reducing the therapeutic arsenal available for treatment of infectious diseases. In the present study, we developed a new class of compounds with antibacterial activity obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service