Merck
  • Thrombosis recanalization by paeoniflorin through the upregulation of urokinase‑type plasminogen activator via the MAPK signaling pathway.
Thrombosis recanalization by paeoniflorin through the upregulation of urokinase‑type plasminogen activator via the MAPK signaling pathway.
Molecular medicine reports (2016-04-16)
Songshan Ye, Bingyu Mao, Lei Yang, Weiyun Fu, Junran Hou
RESUMEN
Paeoniflorin, the major component of Paeonia lactiflora pall, has previously been reported to prevent thrombosis. Plasminogen activator urokinase (uPA) is a serine protease that markedly facilitates normal thrombosis resolution. Paeoniflorin and uPA have been linked to the mitogen‑activated protein kinase (MAPK) signaling pathway. In the current study, the influence of paeoniflorin on the expression of uPA was investigated and the underlying regulatory mechanism was preliminarily determined. The prothrombotic state of the model animals treated with paeoniflorin were assessed by enzyme‑linked immunosorbent assay (ELISA). Additionally, the cytotoxicity of paeoniflorin on human umbilical vein endothelial cell (HUVEC) cultures was estimated using a methyl thiazolyl tetrazolium assay and the possible pathways involved in the interaction between paeoniflorin and uPA were evaluated using western blot analysis. The ELISA results demonstrated that the levels of 6‑keto prostaglandin F1a, fibronectin and uPA were significantly upregulated by treatment with paeoniflorin compared with control (P<0.05). By contrast, the expression of fibrinogen, D‑dimer and thromboxane B2 were inhibited. With an increase in the concentration of paeoniflorin the cell viability of HUVECs decreased gradually. The results of western blot analysis demonstrated that paeoniflorin increased the phosphorylation of MAPK 14 (p38) and MAPK 8 (JNK). The present study demonstrated that paeoniflorin has the potential to improve the prethrombotic state and recanalize thrombosis by increasing the expression of uPA, which may be mediated via regulation of the p38 and JNK MAPK signaling pathways. However, this treatment effect was dependent on the concentration of paeoniflorin used, an unsuitable concentration of the agent would result in a negative effect on the anti‑thrombosis pathways.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Alkaline Phosphatase, Human Placental−Agarose antibody produced in mouse, clone 8B6, purified immunoglobulin, PBS suspension