Merck
SOD3 reduces inflammatory cell migration by regulating adhesion molecule and cytokine expression.
PloS one (2009-06-06)
Juha P Laurila, Lilja E Laatikainen, Maria D Castellone, Mikko O Laukkanen
RESUMEN
Inflammatory cell migration characteristic of ischemic damages has a dual role providing the tissue with factors needed for tissue injury recovery simultaneously causing deleterious development depending on the quality and the quantity of infiltrated cells. Extracellular superoxide dismutase (SOD3) has been shown to have an anti-inflammatory role in ischemic injuries where it increases the recovery process by activating mitogen signal transduction and increasing cell proliferation. However, SOD3 derived effects on inflammatory cytokine and adhesion molecule expression, which would explain reduced inflammation in vascular lesions, has not been properly characterized. In the present work the effect of SOD3 on the inflammatory cell extravasation was studied in vivo in rat hind limb ischemia and mouse peritonitis models by identifying the migrated cells and analyzing SOD3-derived response on inflammatory cytokine and adhesion molecule expression. SOD3 overexpression significantly reduced TNFalpha, IL1alpha, IL6, MIP2, and MCP-1 cytokine and VCAM, ICAM, P-selectin, and E-selectin adhesion molecule expressions in injured tissues. Consequently the mononuclear cell, especially CD68+ monocyte and CD3+ T cell infiltration were significantly decreased whereas granulocyte migration was less affected. According to our data SOD3 has a selective anti-inflammatory role in ischemic damages preventing the migration of reactive oxygen producing monocyte/macrophages, which in excessive amounts could potentially further intensify the tissue injuries therefore suggesting potential for SOD3 in treatment of inflammatory disorders.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Eosin B, Dye content 95 %
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y, SAJ special grade, ≥85.0%
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin B, certified by the BSC, Dye content 90 %