Merck
In vitro cytotoxicity of novel pro-apoptotic agent DM-PIT-1 in PEG-PE-based micelles alone and in combination with TRAIL.
Drug delivery (2009-06-27)
Igor Skidan, Benchun Miao, Ritesh V Thekkedath, Parita Dholakia, Alexei Degterev, Vladimir Torchilin
RESUMEN
The purpose of this study was to develope and characterize a micellar formulations of N-{[(2-hydroxy-5- nitrophenyl)amino]carbonothioyl}-3,5-dimethylbenzamide (DM-PIT-1)-a new small molecule non-lipid antagonist of phopshotidylinositol-3.4.5-triphopshate and inhibitor of the PI3-kinase pathway. Micelle-forming PEG(2000)-PE was used to solubilize DM-PIT-1. To improve the specificity of the micellar DM-PIT-1, cancer-targeting anti-nucleosomal mAb2C5 antibodies as well as Tumor necrosis factor- Related Apoptosis-Inducing Ligand (TRAIL) were attached to the surface of polymeric micelles. DM-PIT-1 was effectively incorporated (> 70%) into 14-16 nm micelles, which had a negative surface zeta potential of 4-5 mV. Micellar DM-PIT-1 demonstrated high in vitro cytotoxicity against various cancer cells. An improved potency of the dual-activity DM-PIT-1/TRAIL combination nanoparticles in inducing death of TRAIL-resistant cancer cells was shown. Efficacy of the TRAIL therapy was enhanced by combining it with the 2C5 antibody cancer-targeted micellar form of DM-PIT-1. In conclusion, DM-PIT-1 micellar preparations can be used for targeted combination therapy against TRAIL-resistant cancers.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
PIP3 Antagonist II, DM-PIT-1, The PIP3 Antagonist II, DM-PIT-1, also referenced under CAS 701947-53-7, controls the biological activity of PIP3.