Key Documents
BUA4815-EP
Buckets for Eppendorf® A-4-81 Rotor
rectangular, capacity 500 mL, pack of 2
About This Item
UNSPSC Code:
41103910
material
rectangular
packaging
pack of 2
manufacturer/tradename
Eppendorf® 022638629 (NA)
Eppendorf® 5810730007 (EU)
technique(s)
centrifugation: suitable
capacity
500 mL
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG