HPLC Analysis of Organic Acids on Discovery® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 5 cm × 4.6 mm I.D., 5 μm particles (504947)
mobile phase
[A] 0.1% TFA in [B] water:methanol; (60:40, A:B)
flow rate
2.0 mL/min
column temp.
20 °C
detector
UV, 254 nm
injection
10 μL
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany