GC Analysis of Industrial Solvents on SUPELCOWAX 10
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.32 mm I.D., 1.0 μm (24211)
oven
35 °C (8 min), 4 °C/min to 130 °C (2 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25 cm/sec
injection
0.5 μL, 200:1 split
liner
split, cup design
sample
59 component neat solvent mixture
Description
Legal Information
null