Merck
Fragrance material review on cinnamyl alcohol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2005-04-07)
C S Letizia, J Cocchiara, J Lalko, A Lapczynski, A M Api
ABSTRACT
A toxicologic and dermatologic review of cinnamyl alcohol when used as a fragrance ingredient is presented.
Sigma-Aldrich
Cinnamyl alcohol, 98%
Sigma-Aldrich
Cinnamyl alcohol, ≥98%, FG