Merck
  • Muscle mTORC1 suppression by IL-6 during cancer cachexia: a role for AMPK.
Muscle mTORC1 suppression by IL-6 during cancer cachexia: a role for AMPK.
American journal of physiology. Endocrinology and metabolism (2013-03-28)
James P White, Melissa J Puppa, Song Gao, Shuichi Sato, Stephen L Welle, James A Carson
ABSTRACT
Although catabolic signaling has a well-established role in muscle wasting during cancer cachexia, the suppression of anabolic signaling also warrants further investigation. In cachectic tumor-bearing mice, circulating IL-6 levels are associated with suppressed muscle protein synthesis and mTORC1 signaling. We have found AMPK and IGF-I/insulin signaling, two well-known regulators of the mammalian target of rapamycin (mTOR), are altered with the progression of cachexia. How IL-6 can induce suppression of mTORC1 signaling remains to be established. The purpose of this study was to examine mTOR complex 1 (mTORC1) activation and regulation by IL-6 during cancer cachexia. IL-6 effects on mTOR activation were examined in Apc(Min/+) mouse skeletal muscle and C2C12 myotubes. Systemic IL-6 overexpression in Apc(Min/+) mice produced a dose-dependent suppression of mTOR signaling that corresponded to induction of STAT3 and AMPK phosphorylation. This result was also evident in IL-6-treated myotubes. Basal mTOR activation and mTOR responsiveness to glucose administration were suppressed in cachectic skeletal muscle. However, insulin induction of mTOR activity was maintained in IL-6-treated myotubes. Whereas IL-6 suppression of myotube mTOR activity was rescued by AMPK inhibition, inhibition of STAT3 signaling was not sufficient to rescue IL-6 suppression of mTOR activity. Last, treadmill exercise training was able to prevent IL-6-induced inhibition of mTOR signaling in Apc(Min/+) mice independently of activated STAT. In conclusion, we report dose-dependent suppression of mTOR activity by IL-6 and suppressed mTOR responsiveness to glucose administration in Apc(Min/+) mice. IL-6 suppression of mTOR activity was dependent on AMPK activation and independent of STAT signaling in myotubes.
Sigma-Aldrich
5-Amino-4-imidazolecarboxamide, 95%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution