Merck
  • Thermodynamic study of Cu2+ binding to the DAHK and GHK peptides by isothermal titration calorimetry (ITC) with the weaker competitor glycine.
Thermodynamic study of Cu2+ binding to the DAHK and GHK peptides by isothermal titration calorimetry (ITC) with the weaker competitor glycine.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2011-09-08)
Ana Trapaidze, Christelle Hureau, Wojciech Bal, Mathias Winterhalter, Peter Faller
ABSTRACT
The peptides Asp-Ala-His-Lys (DAHK) and Gly-His-Lys (GHK) are naturally occurring Cu(II)-chelating motifs in human serum and cerebrospinal fluid. Here, the sensitive thermodynamic technique isothermal titration calorimetry was used to study the energetics of Cu(II) binding to DAHK and GHK peptides in the presence of the weaker ligand glycine as a competitor. DAHK and GHK bind Cu(II) predominantly in a 1:1 stoichiometry with conditional dissociation constants [i.e., at pH 7.4, in the absence of the competing chelators glycine and 2-(4-(2-hydroxyethyl)-1-piperazinyl)ethanesulfonic acid buffer] of 2.6 ± 0.4 × 10(-14) M and 7.0 ± 1.0 × 10(-14) M, respectively. Furthermore, the apparent ΔH values were measured and the number of protons released upon Cu(II) binding was determined by performing experiments in different buffers. This allowed us to determine the conditional ΔG, ΔH, and ΔS, i.e., corrected for the contributions of the weaker ligand glycine and the buffer at pH 7.4. We found that the entropic and enthalpic contributions to the Cu(II) binding to GHK and DAHK are distinct, with a enthalpic contribution for GHK. The thermodynamic parameters obtained correspond well to those in the literature obtained by other techniques, suggesting that the use of the weaker ligand glycine as a competitor in isothermal titration calorimetry provides accurate data for Cu(II) binding to high-affinity peptides, which cannot be accurately determined without the use of a competitor ligand.
Sigma-Aldrich
Gly-His-Lys acetate salt, BioXtra, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Gly-His-Lys acetate salt, ≥97% (TLC)