Merck
  • Synthesis, conformational analysis, and biological evaluation of 19-nor-vitamin D3 analogues with A-ring modifications.
Synthesis, conformational analysis, and biological evaluation of 19-nor-vitamin D3 analogues with A-ring modifications.
Journal of medicinal chemistry (2009-09-11)
Laura Sánchez-Abella, Susana Fernández, Annemieke Verstuyf, Lieve Verlinden, Vicente Gotor, Miguel Ferrero
ABSTRACT
We have synthesized several isomers of 19-nor-vitamin D analogues possessing a hydroxy group at C-2 as well as novel derivatives bearing an epoxy substituent at the A-ring. All vitamins were prepared in convergent syntheses utilizing the modified Julia olefination. 1alpha,2alpha,25-Trihydroxy-19-nor-vitamin D(3) (3) and 2beta,3beta-epoxy-1alpha,25-dihydroxy-3-deoxy-19-nor-vitamin D(3) (10), which showed the highest affinity to the vitamin D receptor, displayed the highest potency among the tested compounds to inhibit the proliferation of MCF-7 breast cancer cells.
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)