Merck
Extraocular muscle function is impaired in ryr3-/- mice.
The Journal of general physiology (2019-05-16)
Jan Eckhardt, Christoph Bachmann, Marijana Sekulic-Jablanovic, Volker Enzmann, Ki Ho Park, Jianjie Ma, Hiroshi Takeshima, Francesco Zorzato, Susan Treves
ABSTRACT
Calcium is an ubiquitous second messenger mediating numerous physiological processes, including muscle contraction and neuronal excitability. Ca2+ is stored in the ER/SR and is released into the cytoplasm via the opening of intracellular inositol trisphosphate receptor and ryanodine receptor calcium channels. Whereas in skeletal muscle, isoform 1 of the RYR is the main channel mediating calcium release from the SR leading to muscle contraction, the function of ubiquitously expressed ryanodine receptor 3 (RYR3) is far from clear; it is not known whether RYR3 plays a role in excitation-contraction coupling. We recently reported that human extraocular muscles express high levels of RYR3, suggesting that such muscles may be useful to study the function of this isoform of the Ca2+ channel. In the present investigation, we characterize the visual function of ryr3-/- mice. We observe that ablation of RYR3 affects both mechanical properties and calcium homeostasis in extraocular muscles. These changes significantly impact vision. Our results reveal for the first time an important role for RYR3 in extraocular muscle function.
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution