Merck
Expression and activation of the reprogramming transcription factors.
Biochemical and biophysical research communications (2009-11-11)
Jun Guo, Zi-cheng Li, Ying-hong Feng
ABSTRACT
Recently, a series of exciting reports have revealed that terminally differentiated somatic cells can be reprogrammed to generate induced pluripotent stem (iPS) cells via overexpression of a cocktail of transcription factors such as Oct3, Sox2, Klf4, and c-Myc or Oct3, Sox2, Nanog, and Lin28. Most recently, these iPS cells has been used to generate viable, live-born progeny by tetraploid complementation. Reprogramming of iPS cells inaugurates a new era of biology and medicine, it inevitably brings new challenges, e.g., how these factors induce reprogramming and how their expression is regulated. To facilitate iPS cell research, this review focuses on how expression and activation of these transcription factors are regulated.
Sigma-Aldrich
Sox2 TAT human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Sigma-Aldrich
Sox2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture