Skip to Content
Merck
Tetrahydrofurfuryl acetate 97%

Tetrahydrofurfuryl acetate

Tetrahydrofurfuryl acetate 97%
Synonym(s):
Tetrahydro-2-furylmethyl acetate
Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing