
Spectinomycin Sulfate

Synonym(s):
Spectinomycin sulfate tetrahydrate
Empirical Formula (Hill Notation):
C14H26N2O11S · 4H2O
CAS Number:
Molecular Weight:
502.49
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing