
Retigabine

Synonym(s):
D-23129, Ezogabine, N-(2-Amino-4-(4-fluorobenzylamino)phenyl)carbamic acid ethyl ester
Empirical Formula (Hill Notation):
C16H18FN3O2
CAS Number:
Molecular Weight:
303.33
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing