
Pseudoephedrine sulfate

Synonym(s):
(1S,2S)-(+)-Pseudoephedrine sulfate salt, α-(1-Methylaminoethyl)benzyl alcohol sulfate
Empirical Formula (Hill Notation):
C20H32N2O6S
CAS Number:
Molecular Weight:
428.54
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing