
Powdered milk thistle extract

Synonym(s):
Silybum marianum extract, Milk thistle extract
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing