
Polyoxyl 4 lauryl ether

Synonym(s):
Tetraethylene glycol monododecyl ether, C12E4, Dodecyl tetraethylene glycol ether, Dodecyltetraglycol
Linear Formula:
CH3(CH2)11(OCH2CH2)4OH
CAS Number:
Molecular Weight:
362.54
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing