
Oxolinic acid

Synonym(s):
5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid
Empirical Formula (Hill Notation):
C13H11NO5
CAS Number:
Molecular Weight:
261.23
Products
Product Category
Brand
Formula Weight
Manufacturer
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing