
Oxazepam Related Compound C

Synonym(s):
6-Chloro-4-phenylquinazoline-2-carbaldehyde, 6-Chloro-4-phenylquinazoline-2-carboxaldehyde
Empirical Formula (Hill Notation):
C15H9ClN2O
CAS Number:
Molecular Weight:
268.70
Products
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2