Skip to Content
Merck
Methyl eugenol analytical standard

Methyl eugenol

Methyl eugenol analytical standard
Synonym(s):
Eugenol methyl ether, 4-Allyl-1,2-dimethoxybenzene, Eugenyl methyl ether
Linear Formula:
H2C=CHCH2C6H3(OCH3)2
CAS Number:
Molecular Weight:
178.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing