
Laminin

Synonym(s):
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane
CAS Number:
Products
Product Category
Biological Source
Gene Alias
Manufacturer
Physical Form
Quality Segment
Sterility
Sterilization Method
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing