
Isobutyl isobutyrate

Synonym(s):
2-Methylpropyl 2-methylpropanoate
Linear Formula:
(CH3)2CHCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
144.21
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing