
Human chorionic gonadotropin

Synonym(s):
Chorionic gonadotropin human, Choriogonin, HCG
CAS Number:
Products
Product Category
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing