
Galactose, plant-derived

Synonym(s):
D-(+)-Galactose, Galactose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Products
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing