
Flunarizine for system suitability

Synonym(s):
Flunarizine dihydrochloride, 1-[Bis(4-fluorophenyl)methyl]-4-(3-phenyl-2-propenyl)piperazine dihydrochloride
Empirical Formula (Hill Notation):
C26H26F2N2 · 2HCl
CAS Number:
Molecular Weight:
477.42
Products
Brand
Formula Weight
Manufacturer
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing