
Ethiofencarb

Empirical Formula (Hill Notation):
C11H15NO2S
CAS Number:
Molecular Weight:
225.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing