
Esomeprazole magnesium trihydrate

Synonym(s):
(S)-Omeprazole magnesium trihydrate
Empirical Formula (Hill Notation):
C34H36MgN6O6S2 · 3H2O
CAS Number:
Molecular Weight:
767.17
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing