
D-Fructose 6-phosphate disodium salt hydrate

Empirical Formula (Hill Notation):
C6H11Na2O9P · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing