
Chlorphenamine Impurity D

Synonym(s):
α-(4-Chlorophenyl)-α-[2-(dimethylamino)ethyl]-2-pyridineacetonitrile hydrochloride, (2RS)-2-(4-Chlorophenyl)-4-dimethylamino-2-(2-pyridyl)butyronitril hydrochloride, Chlorpheniramine nitrile hydrochloride
Empirical Formula (Hill Notation):
C17H18ClN3 · HCl
Molecular Weight:
336.26
Products
Brand
Formula Weight
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing