
Basic butylated methacrylate copolyme

Synonym(s):
2-Methyl-2-propenoic acid butyl ester polymer with 2-(dimethylamino)ethyl 2-methyl-2-propenoate and methyl 2-methyl-2-propenoate
CAS Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing