
Astilbin

Synonym(s):
Dihydroquercetin 3-rhamnoside, Taxifolin 3-rhamnoside
Empirical Formula (Hill Notation):
C21H22O11
CAS Number:
Molecular Weight:
450.39
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing