
Albendazole for system suitability

Synonym(s):
Albendazole, Methyl 5-(propylthio)-2-benzimidazolecarbamate
Empirical Formula (Hill Notation):
C12H15N3O2S
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing