
Acridine orange zinc chloride double salt (C.I.46005)

Empirical Formula (Hill Notation):
C17H20Cl3N3Zn
CAS Number:
Molecular Weight:
438.11
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing